Learn More
Stanbio™ RaPET™ ASO Test
SDP
Catalog No. 23022326 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Quantity:
50 Tests
Catalog No. Quantity
23-022326 50 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-022326 Supplier Stanbio™ Supplier No. 1125050
This item is not returnable. View return policy
Includes
Slides, pipet/mixers, and controls to assure accurate test results, along with an easy-to-use worksheet for documenting semi-quantitative results
Rapid particle enhanced technology provides results in two minutes
  • “No Dilution” technology
  • Color-coded reagents and dropper caps help to reduce chance of errors
  • Utilize semi-quantitative testing procedure for the added convenience of using the same patient serial dilution for all RaPET assays
Specifications
Type RaPET™ ASO Test
Includes Slides (six-well), Latex reagent, positive and negative control and package insert, pipet/mixers, color coded reagents, dropper caps, easy-to-use worksheet, reactive, weakly reactive and non-reactive control along with pipettes/stirrers, dispensing needle, dispensing bottle and 18mm circle test cards
Quantity 50 Tests
Test Time 2 min.