Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ Ser-T-Cal™ Multi-Calibrator Serum
SDP

Catalog No. 22022602 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Quantity:
6 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-022-602 6 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-022-602 Supplier Stanbio™ Supplier No. 0550605

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Multi-calibrator serum to calibrate different chemistry reagent used in manual and automated clinical chemistry methods

  • Stable, lyophilized product prepared from human serum containing additives to provide the defined assay values
  • Store unopened at 2°C to 8°C and stable until expiration date
  • For each constituent, assigned values were obtained using Stanbio test reagents (Differences in chemistry methodologies and manufacture may result in different expected ranges)

Specifications

Content And Storage 2°C to 8°C
For Use With (Equipment) Automated chemistry system
Type Ser-T-Cal™ Multi-Calibrator
Includes 16 Constituents including total and direct bilirubin
Intended Use In-vitro diagnostic use only
Quantity 6 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.