Learn More
Stanbio™ Ser-T-Fy™ I/II Control Assayed Serum
SDP
Catalog No. 22022600 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Type:
Ser-T-Fy I Normal Control Serum
Ser-T-Fy II Abnormal Control Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
22-022-600 Ser-T-Fy I Normal Control Serum
22-022-601 Ser-T-Fy II Abnormal Control Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-022-600 Supplier Stanbio™ Supplier No. G42786
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
A control serum to monitor the accuracy and precision of manual and automated, clinical chemistry methods
  • Offers a stable, lyophilized product prepared from human serum with added chemicals
  • Over 60 different constituents including therapeutic drugs
  • For each constituent, assigned values were obtained with Stanbio test reagents; differences in chemistry methodologies and manufacture may result in different expected ranges
  • Reported with Instrument Specific Values
Specifications
Type Ser-T-Fy I Normal Control Serum
Includes Therapeutic drugs
Intended Use In-vitro diagnostic use only
Quantity 6 x 5 mL