Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ True™ 20 Plus hCG Urine Pregnancy Test Set
SDP

Catalog No. 22022519 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-022-519 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-022-519 Supplier Stanbio™ Supplier No. 1430050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detects 20 mIU/mL hCG for urine samples

  • Only two drops of urine or serum required
  • Provides fast results

Certifications

CLIA waived

Specifications

Accuracy >99%
Content And Storage Room Temperature
Clia Complexity Waived
Detectable Analytes hCG
Sample Type Serum, Urine
Sensitivity 20 mIU/mL
Type hCG Pregnancy Test Kit
Quantity 50 Tests
Specificity >99%
Test Time 10 sec.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.