Learn More
MSC Stanco Banded Waist-Style Ike Jacket, Nomex, 6.0 oz., 1 ea
Catalog No. 5000796890
Encompass
The Stanco Banded Waist-Style Ike Jacket is a flame-resistant protective garment designed for industrial environments. It features a durable Nomex construction, offering safety and comfort.
  • Made from 6.0 oz. Nomex
  • Banded waist style
  • Two slant front pockets
  • Zipper front closure
  • Includes a zipper for a liner (liner not included)
Catalog No. 50-007-96890 Supplier MSC Supplier No. 03747789
The Stanco Banded Waist-Style Ike Jacket is a flame-resistant protective garment designed for industrial environments. It features a durable Nomex construction, offering safety and comfort.
  • Made from 6.0 oz. Nomex
  • Banded waist style
  • Two slant front pockets
  • Zipper front closure
  • Includes a zipper for a liner (liner not included)