Learn More
MSC Stanco Coverall, Flame-Resistant, Nomex IIIA, CAT 1, 1 ea
Catalog No. 5000687207
Encompass
This flame-resistant coverall is designed for protection in hazardous environments, offering robust safety features and durable construction.
  • Elasticized waistband
  • Full action back
  • Two-way concealed brass zipper
  • Two front, rear, chest, and pass-through pockets for extra storage
  • Tool pocket on leg
  • Arc rating: 4.8 cal/cm²
  • Made with Nomex® IIIA fabric
  • Hazard risk category: CAT 1
  • Fabric weight: 4.5 oz/yd²
Catalog No. 50-006-87207 Supplier MSC Supplier No. 03746856
This flame-resistant coverall is designed for protection in hazardous environments, offering robust safety features and durable construction.
  • Elasticized waistband
  • Full action back
  • Two-way concealed brass zipper
  • Two front, rear, chest, and pass-through pockets for extra storage
  • Tool pocket on leg
  • Arc rating: 4.8 cal/cm²
  • Made with Nomex® IIIA fabric
  • Hazard risk category: CAT 1
  • Fabric weight: 4.5 oz/yd²