Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Stanco Coveralls, Indura Ultra Soft, X-Large, Navy, 1 ea

Catalog No. 5000795990
Encompass

These coveralls are designed for protection in hazardous environments. Made from Indura Ultra Soft material, they offer flame-resistant and flame-retardant properties, suitable for industrial applications requiring safety compliance.

  • Garment style: coveralls
  • Garment type: flame-resistant and flame-retardant
  • Size: X-Large
  • Color: navy blue
  • Material: Indura Ultra Soft
  • Hazardous protection level: 2
  • Cuff style: snaps
  • Ankle style: elastic
  • Seam style: safety stitched
  • Features: elastic waist, 2-way concealed zipper, full action back, and elastic waistband
  • Closure type: zipper
  • Seven pockets for storage
  • Country of origin: Mexico

Catalog No. 50-007-95990 Supplier MSC Supplier No. 03747292
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These coveralls are designed for protection in hazardous environments. Made from Indura Ultra Soft material, they offer flame-resistant and flame-retardant properties, suitable for industrial applications requiring safety compliance.

  • Garment style: coveralls
  • Garment type: flame-resistant and flame-retardant
  • Size: X-Large
  • Color: navy blue
  • Material: Indura Ultra Soft
  • Hazardous protection level: 2
  • Cuff style: snaps
  • Ankle style: elastic
  • Seam style: safety stitched
  • Features: elastic waist, 2-way concealed zipper, full action back, and elastic waistband
  • Closure type: zipper
  • Seven pockets for storage
  • Country of origin: Mexico

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.