Learn More
MSC Stanco Ike Jacket, 6.0 oz Nomex, Zipper Closure, 1 ea
Catalog No. 5000686895
Encompass
This banded waist-style Ike jacket is constructed from 6.0 oz. Nomex IIIA Aramid, an inherently flame-resistant material designed to maintain its protective properties through washing and wear.
  • Banded waist-style design
  • Two slant front pockets
  • Zipper front closure
  • Zipper for optional liner (liner not included)
Catalog No. 50-006-86895 Supplier MSC Supplier No. 03747797
This banded waist-style Ike jacket is constructed from 6.0 oz. Nomex IIIA Aramid, an inherently flame-resistant material designed to maintain its protective properties through washing and wear.
  • Banded waist-style design
  • Two slant front pockets
  • Zipper front closure
  • Zipper for optional liner (liner not included)