Learn More
MSC Stanco Nomex IIIA Coverall, 4.5 oz, XL, 1 ea
Catalog No. 5000685512
Encompass
Stanco Nomex® IIIA Aramid Coveralls are flame resistant for the life of the garment. They are constructed with a two-way brass zipper front closure, a by-swing back, and two front, rear, back, and breast pockets with concealed snap closures. The coveralls feature an elastic waistband and five-thread safety stitch.
  • Elasticized waistband
  • Full action back
  • Two-way concealed brass zipper
  • Two front, rear, chest and pass-through pockets for extra storage
  • Tool pocket on leg
  • Fabric: Nomex® IIIA, 4.5 oz/yd²
  • Arc rating: 4.8 cal/cm² (Hazard Risk Category CAT 1)
Catalog No. 50-006-85512 Supplier MSC Supplier No. 03746922
Stanco Nomex® IIIA Aramid Coveralls are flame resistant for the life of the garment. They are constructed with a two-way brass zipper front closure, a by-swing back, and two front, rear, back, and breast pockets with concealed snap closures. The coveralls feature an elastic waistband and five-thread safety stitch.
  • Elasticized waistband
  • Full action back
  • Two-way concealed brass zipper
  • Two front, rear, chest and pass-through pockets for extra storage
  • Tool pocket on leg
  • Fabric: Nomex® IIIA, 4.5 oz/yd²
  • Arc rating: 4.8 cal/cm² (Hazard Risk Category CAT 1)