Learn More
MSC Stanco Safety Arc Flash Hood, Universal, HRC 3, 1 ea
Catalog No. 5000684443
Encompass
This arc flash hood provides critical protection for personnel working in environments with potential arc flash hazards.
  • Universal size for versatile use
  • Hazard Risk Category 3 for robust protection
  • Provides maximum arc flash protection of 25 cal/cm²
Catalog No. 50-006-84443 Supplier MSC Supplier No. 77987956
This arc flash hood provides critical protection for personnel working in environments with potential arc flash hazards.
  • Universal size for versatile use
  • Hazard Risk Category 3 for robust protection
  • Provides maximum arc flash protection of 25 cal/cm²