Stanco Safety Products Indura Ultra Soft HRC2 Coverall is designed for superior protection against electric arc and flash fire exposure. Constructed from flame-resistant cotton and nylon fabrics, it offers guaranteed flame resistance for the life of the garment while ensuring extreme comfort for the wearer.
- Guaranteed flame resistance for the life of the garment
- Extended garment wear life
- Constructed of 88% cotton and 12% nylon flame resistant fabrics
- Hazard risk category (HRC) 2
- Maximum arc flash protection: 12 cal/cm² (for 9 oz navy fabric)
- Complies with NFPA 70E, NFPA 2112, ASTM F1056, OSHA 29CFR 1910.269
- Two-way, concealed brass zipper closure
- Elasticized waistband and full-action back
- Six pockets (two front, two rear, two pass-through)
- Set-in knit wrist sleeve openings and elastic ankle style