Learn More
MSC Stanco Safety Products Coveralls, 8 cal/Sq cm, Medium, 1 ea
Catalog No. 5000789448
Encompass
Stanco Safety Products flame-resistant coveralls provide protection against arc flash hazards. Made from cotton, these navy blue coveralls feature a zipper closure and seven pockets, suitable for various industrial applications, and offer 8 cal/cm² arc flash protection.
  • Two-way concealed zipper
  • Full action back
  • Elastic waistband
  • Zipper closure
  • Seven pockets
Catalog No. 50-007-89448 Supplier MSC Supplier No. 44225092
Stanco Safety Products flame-resistant coveralls provide protection against arc flash hazards. Made from cotton, these navy blue coveralls feature a zipper closure and seven pockets, suitable for various industrial applications, and offer 8 cal/cm² arc flash protection.
  • Two-way concealed zipper
  • Full action back
  • Elastic waistband
  • Zipper closure
  • Seven pockets