Learn More
MSC Stanco Safety Products Coveralls, Medium, Nomex, HRC 1, 1 ea
Catalog No. 5000769207
Encompass
Stanco Safety Products Nomex Coveralls are designed for protection in hazardous environments. They are flame-resistant and flame-retardant, featuring a zipper closure and seven pockets.
  • Elastic waist
  • Two-way concealed zipper
  • Concealed snap closure
  • Full action back
  • Snap cuffs
  • Elastic ankles
  • Safety stitched seams
  • Seven pockets
Catalog No. 50-007-69207 Supplier MSC Supplier No. 03747029
Stanco Safety Products Nomex Coveralls are designed for protection in hazardous environments. They are flame-resistant and flame-retardant, featuring a zipper closure and seven pockets.
  • Elastic waist
  • Two-way concealed zipper
  • Concealed snap closure
  • Full action back
  • Snap cuffs
  • Elastic ankles
  • Safety stitched seams
  • Seven pockets