Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Stanco Safety Products Coveralls, Nomex, X-Large, 1 ea

Catalog No. 5000802345
Encompass

These coveralls are designed for enhanced protection in hazardous environments. Constructed from Nomex, they offer flame-resistant and flame-retardant properties, making them suitable for industrial applications requiring safety compliance.

  • Garment style: coveralls
  • Garment type: flame-resistant, flame-retardant
  • Size: X-Large
  • Color: navy blue
  • Material: Nomex
  • Hazardous protection level: 1
  • Cuff style: snaps
  • Ankle style: elastic
  • Seam style: safety stitched
  • Features: elastic waist, 2-way concealed zipper, concealed snap closure, full action back, elastic waistband
  • Chest size: 46 to 48 in
  • Closure type: zipper
  • Number of pockets: 7

Catalog No. 50-008-02345 Supplier MSC Supplier No. 03747052
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These coveralls are designed for enhanced protection in hazardous environments. Constructed from Nomex, they offer flame-resistant and flame-retardant properties, making them suitable for industrial applications requiring safety compliance.

  • Garment style: coveralls
  • Garment type: flame-resistant, flame-retardant
  • Size: X-Large
  • Color: navy blue
  • Material: Nomex
  • Hazardous protection level: 1
  • Cuff style: snaps
  • Ankle style: elastic
  • Seam style: safety stitched
  • Features: elastic waist, 2-way concealed zipper, concealed snap closure, full action back, elastic waistband
  • Chest size: 46 to 48 in
  • Closure type: zipper
  • Number of pockets: 7

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.