Learn More
MSC Stanco Safety Products Indura FR Coverall, 1 ea
Catalog No. 5000769543
Encompass
This Indura® Flame Resistant Coverall is designed for protection in environments requiring arc flash and flash fire safety. Made from 9 oz Indura® fabric, it provides reliable flame resistance for demanding applications in industries such as oil and gas.
  • Elasticized waistband
  • Full action back
  • Two chest pockets
  • Two rear pockets
  • Two side pockets with pass-throughs
  • One tool pocket
  • Two-way concealed brass zipper closure
Catalog No. 50-007-69543 Supplier MSC Supplier No. 03747441
This Indura® Flame Resistant Coverall is designed for protection in environments requiring arc flash and flash fire safety. Made from 9 oz Indura® fabric, it provides reliable flame resistance for demanding applications in industries such as oil and gas.
  • Elasticized waistband
  • Full action back
  • Two chest pockets
  • Two rear pockets
  • Two side pockets with pass-throughs
  • One tool pocket
  • Two-way concealed brass zipper closure