Learn More
MSC Stanco Safety Products Indura Proban Ike Jacket, 1 ea
Catalog No. 5000788382
Encompass
The Stanco Safety Products Ike Jacket is a banded waist-style jacket constructed from 9 oz. Indura Proban 100% cotton, offering inherent flame-resistant properties. Designed for demanding work environments, this jacket provides protection and durability.
  • Banded waist-style design
  • Two slant front pockets
  • Zipper front closure
  • Equipped with internal zipper for optional liner
  • Flame-resistant properties guaranteed for garment life with proper laundering
Catalog No. 50-007-88382 Supplier MSC Supplier No. 03747854
The Stanco Safety Products Ike Jacket is a banded waist-style jacket constructed from 9 oz. Indura Proban 100% cotton, offering inherent flame-resistant properties. Designed for demanding work environments, this jacket provides protection and durability.
  • Banded waist-style design
  • Two slant front pockets
  • Zipper front closure
  • Equipped with internal zipper for optional liner
  • Flame-resistant properties guaranteed for garment life with proper laundering