Learn More
MSC Stanco Safety Products Nomex Coveralls, X-Large, Navy Blue, 1 ea
Catalog No. 5000677790
Encompass
These Nomex coveralls are designed for protection in hazardous environments. They are flame-resistant and flame-retardant, featuring a navy blue color and a convenient zipper closure.
  • X-large size
  • Seven pockets
  • Elastic waist and waistband
  • Two-way concealed zipper
  • Concealed snap closure
  • Full action back
  • Snaps cuff style
  • Elastic ankle style
  • Safety stitched seams
  • Hazardous protection level one
  • Country of origin: Mexico
Catalog No. 50-006-77790 Supplier MSC Supplier No. 03746732
These Nomex coveralls are designed for protection in hazardous environments. They are flame-resistant and flame-retardant, featuring a navy blue color and a convenient zipper closure.
  • X-large size
  • Seven pockets
  • Elastic waist and waistband
  • Two-way concealed zipper
  • Concealed snap closure
  • Full action back
  • Snaps cuff style
  • Elastic ankle style
  • Safety stitched seams
  • Hazardous protection level one
  • Country of origin: Mexico