Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Stand

Catalog No. 14380973 Shop All BrandTech Scientific Products
Change view
Click to view available options
Color:
Black/White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-380-973 Black/White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-380-973 Supplier BRANDTECH™ BRAND™ Supplier No. 705230
Add to Cart
Edge
Add to Cart

BRAND™ Stand is an accessory part for use with HandyStep™ Touch and HandyStep™ Touch S.

  • Accessory
  • For use with HandyStep™ Touch and Touch S Electronic Repeating Pipette
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color Black/White
Holds Electronic Pipette
Material Plastic
Positions 1
Shape Horizontal
Stackable No
Sterility Non-sterile
For Use With (Equipment) HandyStep Touch / Touch S
Quantity 1
Product Type Stand
Shelves None
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.