Learn More
Braintree Scientific STAND
SDP
Catalog No. NC9600270
Encompass
Rodent Intubation Stand, this platform provides the perfect stage for intubating mice and rats up to 300 grams. The animal is suspended from the incisor wire on the angled side of the stand. **Sold for research purposes only and not for human use**
Catalog No. NC9600270 Supplier Braintree Scientific Supplier No. RIS100
Rodent Intubation Stand, this platform provides the perfect stage for intubating mice and rats up to 300 grams. The animal is suspended from the incisor wire on the angled side of the stand. **Sold for research purposes only and not for human use**