Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Tuttnauer™ Lab Line EL-PV-G-WS Autoclave
SDP

Catalog No. 13890054 Shop All Heidolph North America Products
Change view
Click to view available options
Capacity (Metric):
85 L
160 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
13-890-054 85 L
01-549-773 160 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 13-890-054 Supplier Heidolph™ Supplier No. 023210680
Add to Cart
Add to Cart
Request a Quote
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Designed for quick heat up times and complete drying, this autoclave includes steam generator, and vacuum pump. No plumbing is required for operation, simply fill the reservoirs and plug it in to start using.

Standard Operating Features:

  • Designed to provide high quality, repeatable performance and accountability for a wide range of applications used in modern laboratories
  • For use with Agar preparation as well as the sterilization of liquids, pipettes, glasses, instruments, and biohazards
  • Equipped with a vacuum pump and a steam generator for all laboratory sterilization needs
  • Compact size allows autoclave to fit on a standard benchtop
  • Immediate Steam (G) and Efficient Air Removal (PV) Steam generator in combination with vacuum (PV G) for complete drying
  • PID (Proportional Integral Differential) pressure control
  • Stores the last 200 cycles in built-in memory
  • Two independent, flexible PT100 temperature sensors prevent boil over of liquids and explosions of bottles
  • Highly efficient drying is achieved by uniformly heating the chamber wall of the benchtop autoclave using a coiled pipe
  • The vacuum pump can be used for post vacuum drying to ensure improved drying of porous loads and hollow instruments such as pipette tips
  • Double locking safety device prevents door from opening while chamber is pressurized
  • Control lock-out switch prevents starting a cycle if door is not properly locked
  • Control system protection prevents door from opening at high pressure and temperature
  • All units include trays and printer
  • Unique design with internal reservoirs allows the unit to operate with or without utility connections to treated water, tap water, and a floor sink
  • Applications: Sterilization of liquids, media, instruments, glassware, bioreactors, clothing, and waste

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance UL, cUL, ASME
Capacity (English) 22 gal.
Capacity (Metric) 85 L
Depth (Metric) Chamber 70 cm
Diameter (English) Chamber 15 in.
Depth (English) Chamber 27.6 in.
Diameter (Metric) Chamber 38 cm
For Use With (Application) Sterilization
For Use With (Equipment) Instruments, glassware or liquids
Length (English) 46.9 in.
Height (English) Exterior 53.8 in.
Height (Metric) Exterior 136.6 cm
Length (Metric) 119 cm
Model 3870 HSG-WS
Product Type Autoclave
Temperature (English) 221°F to 278°F
Temperature (Metric) 105°C to 137°C
Weight (English) 220.5 lb.
Display Multi-color
Weight (Metric) 100 kg
Printer Included
Voltage 230 V
Sterility Non-sterile
Dimensions (L x W x H) 46.5 x 28.3 x 53.7 in.
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.