Learn More
Ace Glass, Inc. Stand, aluminum, 18in, 3.4in hole, holds #50 Ace-Thred column
SDP
Catalog No. 50979488
Encompass
18 H powder-coated aluminum, small universal reactor stand with a 3.4 hole. Stand holds a #50 Ace-Thred column.
Catalog No. 50-979-488 Supplier Ace Glass, Inc. Supplier No. 783750
18 H powder-coated aluminum, small universal reactor stand with a 3.4 hole. Stand holds a #50 Ace-Thred column.