Learn More
Med Vet International Charger Stand 3M For 9681 Surgical Clippers, 9682, 1/Case
Catalog No. 502831685
Encompass_Preferred
The 3M™ surgical clipper charger delivers a fast recharge in three hours to minimize downtime It includes a power cord for dependable charging performance The cord comes with a US/Japan plug type It has overcharge protection to prolong battery service life A charge indicator on the clipper enables quick monitoring of the battery level status Its small, compact size saves space and makes it easy to carry
Catalog No. 50-283-1685 Supplier Med Vet International Supplier No. 836202CS
The 3M™ surgical clipper charger delivers a fast recharge in three hours to minimize downtime It includes a power cord for dependable charging performance The cord comes with a US/Japan plug type It has overcharge protection to prolong battery service life A charge indicator on the clipper enables quick monitoring of the battery level status Its small, compact size saves space and makes it easy to carry