Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Stand Here Maintain Social Distancing 6 ft./2 m Floor Sign (Spanish)

Catalog No. 19129556 Shop All Brady Corporation Products
Change view
Click to view available options
Diameter (English):
8 in.
17 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English)
19-129-556 17 in.
19-129-640 8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-129-556 Supplier Brady™ Supplier No. 170270

Mark a safe distance with this pre-printed floor sign that is designed to be placed on the floor 6 ft. (2 m) apart from another sign to provide a visual indication of safe social distancing.

  • Use in areas where lines may form to provide visual guidance of appropriate spacing
  • Made of durable ToughStripe™ Anti-Skid Polyester that includes a rigid, clear overlaminate, and is selectively covered by anti-skid tread
  • Highly visible glossy material resists harsh chemicals and is easy to apply and remove
  • Anti-skid coating helps prevent slips, trips and falls

Specifications

Material Polyester with Anti-Skid Polyester Overlaminate
Adhesive Type Permanent Rubber
Diameter (English) 17 in.
Mounting Type Self-adhesive
Languages Spanish
Displayed Text PARARSE AQUI MANTENER DISTANCIAMIENTO SOCIAL 6 PIES/2 M
Color Black on Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.