Learn More
Med Vet International Pitch-It IV Stand, Floor Stand, 2-Hook, Three Leg, 30007-012, Each
Catalog No. 503046849
Encompass_Preferred
When no longer needed, it can be discarded into recycling Tripod design for stability Made of sturdy aluminum material Lightwieght for easy portablility Not for use with IV bags exceeding 1200 mL or with a pump
Catalog No. 50-304-6849 Supplier Med Vet International Supplier No. 881335EA
When no longer needed, it can be discarded into recycling Tripod design for stability Made of sturdy aluminum material Lightwieght for easy portablility Not for use with IV bags exceeding 1200 mL or with a pump