Learn More
Ace Glass, Inc. STAND,SCALE-UP SERIES,48in
SDP
Catalog No. 5000854788
Encompass
Stand only, designed for the Scale-Up Series reactors. Stand features: 100ml to 6000ml capacity, lightweight aluminum, adjustable leveling feet and three (3) quick adjustment rod-holding brackets for up to 5/8in (16mm) diameter rods. Base dimensions are 19-1/2 x 24-3/4in (DxW) with code -01 at 48in height and code -02 at 38 inches tall. Call for custom heights. Not included are our 6442 family of mounting clamps nor our 11177 family of ring supports that must be sized to the supported vessel.
Catalog No. 50-008-54788 Supplier Ace Glass, Inc. Supplier No. 1284101
Stand only, designed for the Scale-Up Series reactors. Stand features: 100ml to 6000ml capacity, lightweight aluminum, adjustable leveling feet and three (3) quick adjustment rod-holding brackets for up to 5/8in (16mm) diameter rods. Base dimensions are 19-1/2 x 24-3/4in (DxW) with code -01 at 48in height and code -02 at 38 inches tall. Call for custom heights. Not included are our 6442 family of mounting clamps nor our 11177 family of ring supports that must be sized to the supported vessel.