Learn More
New England Biolabs, Inc. Standard Taq (Mg-free) Reaction Buffer Pack – 6 ml (4 x 1.5 ml)
SDP
Supplier:  New England Biolabs, Inc. B9015S
Encompass
New England Biolabs supplies a unique 10X reaction buffer and a supplemental reagent for use with Taq DNA Polymerase in reaction systems which require a detergent free, low ionic strength buffer and variable magnesium concentration. The Mg-free reaction buffer and supplemental MgCl2 are provided for users that require complete control over Mg2+ concentration to optimize their specific application.
Catalog No. 50-995-038