Learn More
SPEX CERTIPREP LLC 3P Silica at 1,000 g/mL in H2O
Catalog No. NC2218168
Encompass_Preferred
(HPS by Zeptometrix) Silica ICP Single-Component Standard at 1,000 g/mL in H2O. This standard is traceable to NIST SRM 3150. ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited.18 month expiration date.
Catalog No. NC2218168 Supplier SPEX CERTIPREP LLC 3P Supplier No. 1000SIO24F500
(HPS by Zeptometrix) Silica ICP Single-Component Standard at 1,000 g/mL in H2O. This standard is traceable to NIST SRM 3150. ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited.18 month expiration date.