Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Standard Pump Head for High-Performance Precision Tubing L/S 15, Polycarbonate Housing, SS Rotor
Catalog No. NC9619588
Encompass
This Standard pump head accepts high-performance precision tubing¿ choose these pump heads for better pressure generation, suction lift, viscous fluid transfer, and longer tubing life. Select a pump head for one of the following high-performance tubing sizes: L/S 15, L/S 24, L/S 35, L/S 36.
Catalog No. NC9619588 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0701521
This Standard pump head accepts high-performance precision tubing¿ choose these pump heads for better pressure generation, suction lift, viscous fluid transfer, and longer tubing life. Select a pump head for one of the following high-performance tubing sizes: L/S 15, L/S 24, L/S 35, L/S 36.