Learn More
Alkali Scientific Alkali Scientific™ Standard 2" Aluminum Box with 64-cell dividers, Drain Holes
SDP
Catalog No. 503028636
Encompass_Preferred
2″ Aluminum Box for storage of samples or tubes with 4 Drain Holes Aluminum or stainless steel cryovial boxes come with or without cardboard dividers and fit into standard freezer racks. These boxes conform to international standard dimensions and hold 2 to 5 milliliter cryo vials or centrifuge tubes. They can be used as cryo boxes as well as regular storage boxes. 2" Aluminum freezer storage boxes with multiple options for cell divider included. Fits into standard freezer racks. Can be used as cryo boxes as well as regular storage boxes. Has drain holes.  Dimensions (L x W x H) Lid: 5 1/4 x 5 1/4 x 2″ (133 x 133 x 51mm)Box: 5 x 5 x 1 15/16″ (127.7 x 127.7 x 49mm)
Catalog No. 50-302-8636 Supplier Alkali Scientific Supplier No. SB2A64
2″ Aluminum Box for storage of samples or tubes with 4 Drain Holes Aluminum or stainless steel cryovial boxes come with or without cardboard dividers and fit into standard freezer racks. These boxes conform to international standard dimensions and hold 2 to 5 milliliter cryo vials or centrifuge tubes. They can be used as cryo boxes as well as regular storage boxes. 2" Aluminum freezer storage boxes with multiple options for cell divider included. Fits into standard freezer racks. Can be used as cryo boxes as well as regular storage boxes. Has drain holes.  Dimensions (L x W x H) Lid: 5 1/4 x 5 1/4 x 2″ (133 x 133 x 51mm)Box: 5 x 5 x 1 15/16″ (127.7 x 127.7 x 49mm)