Learn More
Bio Rad Laboratories STANDARD 302 NM UV LAMPS
Catalog No. 509010314
Encompass
Lamp, UV; Bio-Rad; Standard; For Molecular Imager Gel Doc XR and ChemiDoc XRS Systems; 302nm; 6
Catalog No. 50-901-0314 Supplier Bio Rad Laboratories Supplier No. 1708097
Lamp, UV; Bio-Rad; Standard; For Molecular Imager Gel Doc XR and ChemiDoc XRS Systems; 302nm; 6