Learn More
Cole-Parmer Myron L 30 QCP (NIST) conductivity calibration standard, 46.7 µS  DFS Item
Manufacturer:  Cole-Parmer 0148991
Standards have NaCl and 442™ conversions included for ease of use. Conductivity values are accurate to within ±1% and are referenced to 25°C. Include a NIST-traceable calibration report supplied by the manufacturer for an additional fee. One 32-oz bottle.
Catalog No. 50-631-768