Learn More
HANNA INSTRUMENTS INC pH 7.01 Calibration Buffer Solution (500 mL)
SDP
Catalog No. NC1828926
Encompass
HI7007L is a premium quality pH 7.01 buffer solution. With lot number and expiration date clearly marked on the label and an air tight, tamper-proof seal to ensure the quality of the solution, Hanna's line of pH calibration solutions are an easy way to get great pH measurements every time.Air tight bottle with tamper-proof seal for quality.Color coded label for easy identification of ph 7 buffer value.Perfect for routine maintenance to ensure accurate readings while extending the life of the electrode.
Catalog No. NC1828926 Supplier HANNA INSTRUMENTS INC Supplier No. HI7007L
HI7007L is a premium quality pH 7.01 buffer solution. With lot number and expiration date clearly marked on the label and an air tight, tamper-proof seal to ensure the quality of the solution, Hanna's line of pH calibration solutions are an easy way to get great pH measurements every time.Air tight bottle with tamper-proof seal for quality.Color coded label for easy identification of ph 7 buffer value.Perfect for routine maintenance to ensure accurate readings while extending the life of the electrode.