Learn More
Cole-Parmer Myron L 700 QCP (NIST) conductivity calibration standard, 700 µS
Catalog No. 50631780 Shop All Cole-Parmer Products
Encompass_Preferred
Standards have NaCl and 442™ conversions included for ease of use. Conductivity values are accurate to within ±1% and are referenced to 25°C. Include a NIST-traceable calibration report supplied by the manufacturer for an additional fee. One 32-oz bottle.
Catalog No. 50-631-780 Supplier Cole-Parmer Supplier No. 0148994
Standards have NaCl and 442™ conversions included for ease of use. Conductivity values are accurate to within ±1% and are referenced to 25°C. Include a NIST-traceable calibration report supplied by the manufacturer for an additional fee. One 32-oz bottle.