Learn More
Analytical Sales & Services Standard 96-Position Radiochemistry Conversion Kit
Catalog No. 5000804415
Encompass
Radiochemistry Conversion Kit\nUse with Para-dox Gen I Reaction Block Vial Rack (96973 or 96960)\n\n• 96 Position\n• Anodized Aluminum\n• for 8x30mm Vials\n• Includes Transfer Plate, Top cover, PFA Film, Cap Mat and Required Hardware (vials not included)\n\nVials are held in place with a novel PFA “webbing” solution. Easily transfer your entire 96-vial array to different stations via the transfer plate. Note: The PFA film/webbing is meant for one time use. Once vials are pushed through the PFA and then later removed, the PFA will be permanently stretched out and deformed.
Catalog No. 50-008-04415 Supplier Analytical Sales & Services Supplier No. 96940
Radiochemistry Conversion Kit\nUse with Para-dox Gen I Reaction Block Vial Rack (96973 or 96960)\n\n• 96 Position\n• Anodized Aluminum\n• for 8x30mm Vials\n• Includes Transfer Plate, Top cover, PFA Film, Cap Mat and Required Hardware (vials not included)\n\nVials are held in place with a novel PFA “webbing” solution. Easily transfer your entire 96-vial array to different stations via the transfer plate. Note: The PFA film/webbing is meant for one time use. Once vials are pushed through the PFA and then later removed, the PFA will be permanently stretched out and deformed.