Learn More
Waters Corp Waters Corporation Pico-Tag Standard, Reference, Amino Acid, Reversed Phase, Optical
Catalog No. 50784991 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Pico-Tag Standard Acidic/Neutral is a reference type standard developed for amino acid applications. It is designed for use with LC systems utilizing reversed phase separation mode and optical detection.
  • Reversed phase separation mode
  • Optical detection method
  • Liquid formulation
  • Compatible with LC systems
  • 1 year shelf life upon reconstitution
  • Reference type standard
  • Suitable for amino acid applications
  • 1 unit per package
  • Contains hazardous material, requiring special freight handling and potentially additional charges
Catalog No. 50-784-991 Supplier Waters Corp Supplier No. WAT010948
The Pico-Tag Standard Acidic/Neutral is a reference type standard developed for amino acid applications. It is designed for use with LC systems utilizing reversed phase separation mode and optical detection.
  • Reversed phase separation mode
  • Optical detection method
  • Liquid formulation
  • Compatible with LC systems
  • 1 year shelf life upon reconstitution
  • Reference type standard
  • Suitable for amino acid applications
  • 1 unit per package
  • Contains hazardous material, requiring special freight handling and potentially additional charges