Learn More
TA Instruments Standard Aluminum Hermetic Pans, Use with Standard Aluminum Hermetic Lids, Hermetic Seal Mode, Tight Vapor Containment, Reliable Thermal Results, Requires Tzero Sample Press or Standard Press, 50 Pack
Catalog No. 5000580909 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments Standard Aluminum Hermetic Pans are for use with Standard Aluminum Hermetic Lids in hermetic seal mode. Sample encapsulation requires either the Tzero Sample Press or Standard Press. These pans are ideal for analysis requiring tight vapor containment and reliable thermal results.
Catalog No. 50-005-80909 Supplier TA Instruments Supplier No. 901685.901
Waters TA Instruments Standard Aluminum Hermetic Pans are for use with Standard Aluminum Hermetic Lids in hermetic seal mode. Sample encapsulation requires either the Tzero Sample Press or Standard Press. These pans are ideal for analysis requiring tight vapor containment and reliable thermal results.