Learn More
TA Instruments Standard Aluminum Lids, Use with Standard and Classic Aluminum Pans, Crimp Seal Mode, Secure Sealing, Maintain Sample Integrity, Requires Tzero Sample Press or Standard Press, DSC Measurements, 200 Pack
Catalog No. 5000580596 Shop All Waters TA Instruments Products
Encompass
Waters TA Instruments Standard Aluminum Lids are for use with Standard Aluminum Pans and Classic Aluminum Pans in crimp seal mode. Sample encapsulation requires either the Tzero Sample Press or Standard Press. They provide secure sealing for DSC measurements and maintain sample integrity.
Catalog No. 50-005-80596 Supplier TA Instruments Supplier No. 901684.901
Waters TA Instruments Standard Aluminum Lids are for use with Standard Aluminum Pans and Classic Aluminum Pans in crimp seal mode. Sample encapsulation requires either the Tzero Sample Press or Standard Press. They provide secure sealing for DSC measurements and maintain sample integrity.