Learn More
TA Instruments Tzero Pans Pkg of 100, Aluminum, Crimp or Hermetic Sealing, Flat Bottoms for Maximum Contact with Tzero Cell Sensors, Crimp Seal Mode with Tzero Lids P/N 901671.901, Hermetic Seal Mode with Tzero Hermetic Lids P/N 901684.901, Sample Encapsulation Requires Tzero Sample Press Kit P/N 901600.901
Catalog No. 5000580593 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Tzero Pans Pkg Of 100 are aluminum pans designed for maximum performance in crimp or hermetic sealing of various sample sizes. They have extremely flat bottoms for maximum contact with Tzero cell sensors. They can be used in crimp seal mode with Tzero Lids P/N 901671.901, or in hermetic seal mode with Tzero Hermetic Lids P/N 901684.901. Sample encapsulation requires the Tzero Sample Press Kit P/N 901600.901.
Catalog No. 50-005-80593 Supplier TA Instruments Supplier No. 901683.901
Waters TA Instruments Tzero Pans Pkg Of 100 are aluminum pans designed for maximum performance in crimp or hermetic sealing of various sample sizes. They have extremely flat bottoms for maximum contact with Tzero cell sensors. They can be used in crimp seal mode with Tzero Lids P/N 901671.901, or in hermetic seal mode with Tzero Hermetic Lids P/N 901684.901. Sample encapsulation requires the Tzero Sample Press Kit P/N 901600.901.