Learn More
Biosurface Technologies Corp Standard Biofilm Annular Reactor, Mid- to High-Shear Laboratory Biofilm Growth and Monitoring Device, BioSurface Technologies Corporation.
Catalog No. NC3662416
Encompass
"Evaluation of surface-associated biological reactions require surface-specific evaluations. By matching surface shear and transport conditions within the Biofilm Annular Reactor to your process conditions, relevant evaluations of your water treatment processes are possible.The BST Model 1420 Biofilm Annular Reactor consists of a stationary outer cylinder and a rotating inner cylinder. Process fluid is circulated in the annulus between the two cylinders. This model operates with a variable-speed motor. The rotational speed of the inner cylinder is set to provide liquid-surface shear similar to the pipe flow shear of the process water system you are evaluating.The Biofilm Annular Reactor (BAR 1420 LS) is manufactured using an inner, slotted polycarbonate cylinder and a glass outer cylinder. Twenty (20) slides are flush-mounted on the rotating inner cylinder. The slides are available in a variety of metals and plastics.
Catalog No. NC3662416 Supplier Biosurface Technologies Corp Supplier No. BAR 1420 LS
Add to Cart
Add to Cart
Request a Quote
"Evaluation of surface-associated biological reactions require surface-specific evaluations. By matching surface shear and transport conditions within the Biofilm Annular Reactor to your process conditions, relevant evaluations of your water treatment processes are possible.The BST Model 1420 Biofilm Annular Reactor consists of a stationary outer cylinder and a rotating inner cylinder. Process fluid is circulated in the annulus between the two cylinders. This model operates with a variable-speed motor. The rotational speed of the inner cylinder is set to provide liquid-surface shear similar to the pipe flow shear of the process water system you are evaluating.The Biofilm Annular Reactor (BAR 1420 LS) is manufactured using an inner, slotted polycarbonate cylinder and a glass outer cylinder. Twenty (20) slides are flush-mounted on the rotating inner cylinder. The slides are available in a variety of metals and plastics.