Learn More
Thermo Scientific Standard tube dispensing cassette, 1/each
Supplier:  Thermo Scientific 24072670
Encompass_Preferred
Dispenser; Automated; Microplate; Accessory; 8-channel; autoclavable; Standard tube dispensing cassette; For volumes 5-2500uL; For Use With: Multidrop Combi, Multidrop 384 and Multidrop DW
Catalog No. 50-950-372