Learn More
Ace Glass, Inc. Standard 4in conical flange clamp complete
SDP
Catalog No. 509557324
Encompass
Anodized aluminum clamp is designed to fit 4" (100mm) conical flanges and slides open to accomm ODate flasks. Includes convenient 1/2" x 8" removable arm for attaching to racks, 3 SS swing bolts, and 4 silicone liners for operation to 500F.
Catalog No. 50-955-7324 Supplier Ace Glass, Inc. Supplier No. 649610
Anodized aluminum clamp is designed to fit 4" (100mm) conical flanges and slides open to accomm ODate flasks. Includes convenient 1/2" x 8" removable arm for attaching to racks, 3 SS swing bolts, and 4 silicone liners for operation to 500F.