Learn More
TA Instruments Standard Coated Aluminum Hermetic Lids, Use with Standard Coated Aluminum Hermetic Pans, Hermetic Seal Mode, Aqueous or Organic Liquids, Requires Tzero Sample Press or Standard Press, 200 Pack
Catalog No. 5000580766 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Standard Coated Aluminum Hermetic Lids are for use with Standard Coated Aluminum Hermetic Pans in hermetic seal mode for analysis of aqueous or organic liquids. Sample encapsulation requires either the Tzero Sample Press or Standard Press.
Catalog No. 50-005-80766 Supplier TA Instruments Supplier No. 900786.901
Waters TA Instruments Standard Coated Aluminum Hermetic Lids are for use with Standard Coated Aluminum Hermetic Pans in hermetic seal mode for analysis of aqueous or organic liquids. Sample encapsulation requires either the Tzero Sample Press or Standard Press.