Learn More
TA Instruments Standard Coated Aluminum Hermetic Pans, Aqueous or Organic Liquids, Hermetic Seal Mode, Requires Standard Coated Aluminum Hermetic Lids, Use with Tzero Sample Press or Standard Press, 200 Pack
Catalog No. 5000580809 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments Standard Coated Aluminum Hermetic Pans are used for analysis of aqueous or organic liquids in hermetic seal mode. Requires Standard Coated Aluminum Hermetic Lids. Sample encapsulation requires either the Tzero Sample Press or Standard Press.
Catalog No. 50-005-80809 Supplier TA Instruments Supplier No. 900760.901
Waters TA Instruments Standard Coated Aluminum Hermetic Pans are used for analysis of aqueous or organic liquids in hermetic seal mode. Requires Standard Coated Aluminum Hermetic Lids. Sample encapsulation requires either the Tzero Sample Press or Standard Press.