Learn More
Andwin Scientific STANDARD COMB FOR DNA PLUS 12
SDP
Catalog No. 50820248
Encompass_Preferred
Standard comb for DNA Plus;12 x 14 cm Mini Agarose Gel System. Teeth/Comb: 20. Thickness: 1.5 mm. Tooth Width: 4.5 mm. Volume/Well: 30 ul.
Catalog No. 50-820-248 Supplier Andwin Scientific Supplier No. 34875520
Standard comb for DNA Plus;12 x 14 cm Mini Agarose Gel System. Teeth/Comb: 20. Thickness: 1.5 mm. Tooth Width: 4.5 mm. Volume/Well: 30 ul.