Learn More
Andwin Scientific STANDARD COMB FOR DNA PLUS 12
SDP
Supplier:  Andwin Scientific 34875524
Encompass_Preferred
Standard comb for DNA Plus;12 x 14 cm Self-Circulating Mini Agarose Gel System. Teeth/Comb: 24. Thickness: 1.5 mm. Tooth Width: 3.0 mm. Volume/Well: 20 ul.
Catalog No. 50-820-249