Learn More
Andwin Scientific STANDARD COMB FOR DNA PLUS 9
SDP
Supplier:  Andwin Scientific 34870510
Encompass_Preferred
Standard comb for DNA Plus;9 x 11 cm Mini Agarose Gel System. Tooth/Comb: 10. Thickness: 1.5 mm. Tooth Width: 7 mm. Volume/Well: 50 ul.
Catalog No. 50-820-238