Learn More
Andwin Scientific STANDARD COMB FOR DNA PLUS 9
SDP
Catalog No. 50820239
Encompass_Preferred
Standard comb for DNA Plus;9 x 11 cm Mini Agarose Gel System. Tooth/Comb: 14. Thickness: 1.5 mm. Tooth Width: 4 mm. Volume/Well: 30 ul.
Catalog No. 50-820-239 Supplier Andwin Scientific Supplier No. 34870514
Standard comb for DNA Plus;9 x 11 cm Mini Agarose Gel System. Tooth/Comb: 14. Thickness: 1.5 mm. Tooth Width: 4 mm. Volume/Well: 30 ul.