Learn More
Andwin Scientific STANDARD COMB FOR DNA PLUS ST
SDP
Catalog No. 50820208
Encompass_Preferred
Standard comb for DNA Plus;Standard Horizontal Gel System. Teeth/Comb: 36. Thickness: 1.5 mm. Tooth Width: 3.5 mm. Volume/Well: 25 ul.
Catalog No. 50-820-208 Supplier Andwin Scientific Supplier No. 34820536
Standard comb for DNA Plus;Standard Horizontal Gel System. Teeth/Comb: 36. Thickness: 1.5 mm. Tooth Width: 3.5 mm. Volume/Well: 25 ul.