Learn More
Hoefer, Inc Standard Combs for SE660, SE600X Chroma, SE600, SE640, SE410, and SE400
SDP
Supplier:  Hoefer, Inc SE511281.0
Encompass_Preferred
Standard Comb, 28 Wells, 1 mm Thickness, 2.7 Width, 2.7 Volume. Combs are precision machined from PTFE which is highly resistant to acids, bases, and all commonly used buffer systems. An important benefit of PTFE combs, compared to other types, is that they are easily removed while maintaining gel integrity. They are lightweight and virtually unbreakable when used with reasonable care.
Catalog No. NC2957450