Learn More
Andwin Scientific Single-Use, 100 mL (3.4 oz.) Polyethylene Bottles, 10 uS, 6/pk
SDP
Catalog No. NC1255361
Encompass_Preferred
These Certified Reference Materials (CRM) calibrate conductivity meters and probes for maximum accuracy. They are compatible with all makes of equipment, and can be used in lab or field.
Catalog No. NC1255361 Supplier Andwin Scientific Supplier No. 23226650
These Certified Reference Materials (CRM) calibrate conductivity meters and probes for maximum accuracy. They are compatible with all makes of equipment, and can be used in lab or field.